Essentials Double-Ended Spoons, 304 Stainless Steel, Antylia Scientific
Supplier: Antyila Scientific
06700-70 06700-71 06700-72 06700-73 06700-74
76533-778EA 14.05 USD
76533-778 76533-780 76533-782 76533-784 76533-786
Essentials Double-Ended Spoons, 304 Stainless Steel, Antylia Scientific
Spoons
Take different sample sizes from the same spoon.
  • 304 stainless steel (SS) construction for excellent chemical compatibility
  • BSE/TSE free
  • Non sterile
Simply flip around the spoon to take different sample sizes. Spoons are constructed of 304 SS with a polished smooth finish—ideal for powders and liquid.
Certifications: Supplier Certificate of Conformity is included with product shipment.