Cole-Parmer® Essentials Color-Coded Cable/Zip Ties
Supplier: Antyila Scientific
06842-29 06842-30 06842-31 06842-32 06830-31 06830-32 06830-33 06830-61 06830-62 06842-33 06842-34 06842-35 06842-36 06842-37 06842-38 06842-39 06842-40 06842-41 06830-41 06830-63 06830-64 06830-65
76579-818CS 1.96 USD
76579-818 76579-820 76579-896 76579-872 76579-828 76579-946 76579-906 76579-974 76579-930 76579-868 76579-976 76579-938 76579-904 76579-844 76579-978 76579-810 76579-816 76579-900 76579-966 76579-934 76579-940 76579-874
Cole-Parmer® Essentials Color-Coded Cable/Zip Ties
Ties
These ties are injection-molded of ⁶/₆ nylon, and won’t corrode or stretch.
  • Ties hold securely, even in vibrating, oily, or wet environments
  • Easily apply ties by hand or use a tensioning tool
Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2.
Ties withstand temperatures to 85 °C (185 °F ) and have a melting point of 257 °C (495 °F). All ties remain flexible to −40 °C (−104 °F). Efficient range of ties may be higher or lower depending upon specific applications.
Ordering information: Tools must be ordered separately.