Cole-Parmer® Essentials Mountable Cable Ties
Supplier: Antyila Scientific
06842-53 06842-54 06842-55 06842-56 06842-57 06842-58 06842-51 06842-52
76579-830CS 9.42 USD
76579-830 76579-922 76579-914 76579-866 76579-894 76579-916 76579-878 76579-892
Cole-Parmer® Essentials Mountable Cable Ties
Ties
Mountable cable ties offer two products in one. Easily bundle cables together and permanently screw mount the ties to a wall, panel, or other surfaces using the integrated mounting eyelet on the cable tie head.
  • Requires no additional clips or mounts
  • Self-lock feature securely bundles wires and cables keeping them neat and organized
Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2. Ties withstand temperatures to 85 °C (185 °F ) and have a melting point of 257 °C (495 °F).