FrameStar® PCR Plates, 96-Well, Fully Skirted, Low Profile, Azenta Life Sciences
Supplier: Azenta Life Sciences
FrameStar®
4TI-LB0960/RIG 4TI-0960/C 4TI-0961 4TI-0960/X 4TI-OX960/C/SBC 4TI-0966 4TI-0960/G 4TI-0960/B 4TI-0960/R 4TI-0961/C 4TI-0960/W 4TI-0960 4TI-0960/RIG
76619-082CS 359.83 USD
76619-082 76629-106 76629-126 76628-988 76629-102 76629-036 76638-164 76629-114 76629-012 76629-032 76629-000 76619-076 76619-068
FrameStar® PCR Plates, 96-Well, Fully Skirted, Low Profile, Azenta Life Sciences
Microplates PCR Plates
Low profile, 0.1 ml polypropylene wells, polycarbonate frame, cut corner H1; working volume: <100 µl, total well capacity: 200 µl. Ideal for use with robotic systems. Compatible with standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for Improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
The low profile wells of this plate are shorter than 'standard' profile wells, which decrease the 'dead space' between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction.
This is especially recommended for reaction volumes below 20 µl. The rigid polycarbonate skirt of this design eliminates warping and distortion during PCR, and makes it ideal for use with robotic systems.
Ordering information: Ultra-low DNA binding option available. Available barcoded upon request.
Packaging: 50 plates per case.