SingleShot Buffer Pouches

Supplier: NuSep
DBM-TBE-01 DBM-PBS-01 DBM-SSC-01 DBM-LBM-01 DBM-TGS-01 DBM-LBA-01 DBM-TRG-01 DBM-TAE-01
470348-096EA 62.3 USD
470348-096 470348-100 470348-110 470348-098 470348-104 470348-102 470348-108 470348-106
SingleShot Buffer Pouches
Buffers Biological Buffers
Each pouch contains a convenient, pre-mixed buffer solution; just add water!

  • Quality – molecular biology-grade buffers of highest purity
  • Convenient – pre-measured powder in sealed pouches
  • Fast – dissolve and use right away
  • Standardized – guaranteed reproducible results for your electrophoresis gels and Western blots

NuSep’s SingleShot Buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-measured molecular biology-grade chemicals in 500 ml of deionized water to obtain a ready-to-use 1x buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.

Buffer Selections:
TGS (Tris-Glycine SDS) – Gel Running Buffer-Cat# DBM-TGS-01
PBS (Phosphate Buffer Saline) – Western Blotting-Cat# DBM-PBS-01
TG (Tris-Glycine) – Transfer Buffer-Cat# DBM-TRG-01
TBE (Tris-Boric EDTA Buffer) – DNA/RNA Electrophoresis Buffer-Cat# DBM-TBE-01
TAE (Tris-Acetate EDTA Buffer) – DNA/RNA Electrophoresis Buffer-Cat# DBM-TAE-01
SSC (Saline Sodium Citrate Buffer) – Southern and Northern Hybridization-Cat# DBM-SSC-01

Each packet quickly yields 500 ml of ready-to-use 1x concentration buffer

NuSep also provide LB Agar and LB Media formulations. Buffers are premixed and packaged for your convenience. Just add water and use.

LB Agar (Cat# DBM-LBA-01): Each packet quickly yields 50 ml of ready-to-use 1x concentration working solution.
LB Media (Cat# DBM-LBM-01): Each packet quickly yields 100 ml of ready-to-use 1x concentration working solution.

Delivery information: 20 Buffer Pouches/Box
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR