Magnesium fluoride, powder

Supplier: APOLLO SCIENTIFIC
Warning

PC5020-100G PC5020-25G
76603-976EA 297 USD
76603-976 76607-488
Magnesium fluoride, powder
Magnesium fluoride
Formula: MgF₂
MW: 62.3 g/mol
Boiling Pt: 2260
Melting Pt: 1266
Density: 3.148 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR