Perfluoro-2,5,8,11-tetraoxadodecane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluorotriglyme, Perfluoro(triethylene glycol dimethyl ether), 1,1,1,3,3,4,4,6,6,7,7,9,9,10,10,12,12,12-Octadecafluoro-2,5,8,11-tetraoxadodecane

PC9775-1G PC9775-25G PC9775-5G
76610-560EA 29.54 USD
76610-560 76610-562 76610-564
Perfluoro-2,5,8,11-tetraoxadodecane 98%
Perfluoro-2,5,8,11-tetraoxadodecane
Formula: C₈F₁₈O₄
Boiling Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD08458174
CAS Number: 64028-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR