6-Fluoronaphthalen-1-ol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC908525-100MG PC908525-250MG
76610-010EA 80.48 USD
76610-010 76610-012
6-Fluoronaphthalen-1-ol 95+%
6-Fluoronaphthalen-1-ol
Formula: C₁₀H₇FO
MW: 162.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528810
CAS Number: 804498-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR