2,6-Difluoro-4-methoxybenzamide 98%

Supplier: APOLLO SCIENTIFIC

PC8315-10G PC8315-1G PC8315-5G
76609-330EA 269.94 USD
76609-330 76609-332 76609-334
2,6-Difluoro-4-methoxybenzamide 98%
2,6-Difluoro-4-methoxybenzamide
Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD03094497
CAS Number: 125369-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR