2-Fluoro-5-iodobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Fluoro-5-iodophenyl)methanol

PC6376-1G PC6376-250MG PC6376-5G
76603-676EA 45.64 USD
76603-676 76603-678 76603-680
2-Fluoro-5-iodobenzyl alcohol
2-Fluoro-5-iodobenzyl alcohol
Formula: C₇H₆FIO
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD06202824
CAS Number: 438050-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR