2-Bromo-4-chloro-6-fluorobenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC56942-5G PC56942-1G PC56942-250MG
76602-758EA 88.58 USD
76602-758 76602-762 76602-760
2-Bromo-4-chloro-6-fluorobenzaldehyde 95%
2-Bromo-4-chloro-6-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12400895
CAS Number: 1135531-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR