2-(Fluoromethyl)aniline, N-BOC protected 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC56900-1G PC56900-250MG
76614-314EA 536.84 USD
76614-314 76614-316
2-(Fluoromethyl)aniline, N-BOC protected 95%
2-(Fluoromethyl)aniline, N-BOC protected
Formula: C₁₂H₁₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD31664917
CAS Number: 1374329-41-5
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR