2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-3-(bromomethyl)-4-fluorobenzotrifluoride, 2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl bromide

PC56252-1G PC56252-250MG PC56252-5G
76627-102EA 54.38 USD
76627-102 76627-104 76627-106
2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%
2-Bromo-3-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene
Formula: C₈H₄Br₂F₄
Storage Temperature: Ambient
MDL Number: MFCD31580165
CAS Number: 2090283-00-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR