2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC56187-250MG PC56187-1G
76602-368EA 819 USD
76602-368 76602-370
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₂BrF₄N
MW: 268.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31568568
CAS Number: 2091606-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR