4-(Carboxymethyl)-3-fluorobenzeneboronic acid
Supplier: APOLLO SCIENTIFIC
PC53689-1G
PC53689-250MG
76601-840EA
256.7
USD
76601-840
76601-842
4-(Carboxymethyl)-3-fluorobenzeneboronic acid
4-(Carboxymethyl)-3-fluorobenzeneboronic acid
Formula:
C₈H₈BFO₄ Storage Temperature: Ambient |
MDL Number:
MFCD32220289 CAS Number: 1146614-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...