Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate 95%

Supplier: APOLLO SCIENTIFIC

PC53610-5G PC53610-1G PC53610-250MG
76601-808EA 817.98 USD
76601-808 76601-804 76601-806
Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate 95%
Methyl 2,4-dichloro-5-fluoro-3-nitro-benzoate
Formula: C₈H₄Cl₂FNO₄
Storage Temperature: Ambient
MDL Number: MFCD12172978
CAS Number: 1153285-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR