4-Bromo-3-fluoro-2-methylpyridine 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC535034-100MG PC535034-250MG
76601-448EA 132.42 USD
76601-448 76601-450
4-Bromo-3-fluoro-2-methylpyridine 95+%
4-Bromo-3-fluoro-2-methylpyridine
Formula: C₆H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD16609967
CAS Number: 1211583-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR