2-Bromo-5-chloro-4-fluorobenzaldehyde

Supplier: APOLLO SCIENTIFIC

PC53327-1G PC53327-250MG PC53327-5G
76601-042EA 391.36 USD
76601-042 76601-044 76601-046
2-Bromo-5-chloro-4-fluorobenzaldehyde
2-Bromo-5-chloro-4-fluorobenzaldehyde
Formula: C₇H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28733864
CAS Number: 1806838-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR