3-Bromo-4,5-difluorobenzoic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC53246-1G PC53246-25G PC53246-5G
76600-974EA 30.56 USD
76600-974 76600-976 76600-978
3-Bromo-4,5-difluorobenzoic acid 99%
3-Bromo-4,5-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20233291
CAS Number: 1244642-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR