4-(Difluoromethoxy)-3-fluorobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC53100-25G PC53100-5G PC53100-1G
76600-868EA 991.16 USD
76600-868 76600-870 76600-866
4-(Difluoromethoxy)-3-fluorobenzaldehyde
4-(Difluoromethoxy)-3-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11847153
CAS Number: 1214379-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR