2-(Difluoromethoxy)-5-fluorobromobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-1-(difluoromethoxy)-4-fluorobenzene

PC53074-1G PC53074-250MG PC53074-5G
76600-840EA 529.44 USD
76600-840 76600-842 76600-844
2-(Difluoromethoxy)-5-fluorobromobenzene 97%
2-(Difluoromethoxy)-5-fluorobromobenzene
Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09935620
CAS Number: 936249-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR