5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Diamino-5-fluorobenzotrifluoride, 1,2-Diamino-5-fluoro-3-(trifluoromethyl)benzene

PC5169-1G PC5169-250MG
76600-116EA 185.9 USD
76600-116 76600-118
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine
Formula: C₇H₆F₄N₂
Storage Temperature: Ambient
MDL Number: MFCD03094268
CAS Number: 179062-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR