4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-Bromo-1H-benzimidazol-6-yl)sulphur pentafluoride, (4-Bromo-1H-benzimidazol-6-yl)sulfur pentafluoride, 4-Bromo-6-(pentafluorosulphanyl)-1H-benzimidazole, 4-Bromo-6-(pentafluorosulfanyl)-1H-benzimidazole

PC51190-1G PC51190-250MG PC51190-5G
76606-168EA 406.56 USD
76606-168 76604-914 77431-102
4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%
4-Bromo-6-(pentafluorothio)-1H-benzimidazole
Formula: C₇H₄BrF₅N₂S
Storage Temperature: Ambient
MDL Number: MFCD24369706
CAS Number: 1432075-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR