2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC50370-1G PC50370-250MG
76606-218EA 284.2 USD
76606-218 76607-140
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95+%
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formula: C₉H₇F₆NO
Storage Temperature: Refrigerator
MDL Number: MFCD17015636
CAS Number: 2402-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR