2-Fluoro-4-(methylsulfonyl)benzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC501373-1G PC501373-5G
76606-892EA 92.7 USD
76606-892 76607-202
2-Fluoro-4-(methylsulfonyl)benzaldehyde
2-Fluoro-4-(methylsulfonyl)benzaldehyde
Formula: C₈H₇FO₃S
Storage Temperature: Ambient
MDL Number: MFCD13191594
CAS Number: 1197193-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR