2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Bromo-4-chloro-6-fluorophenyl)methanol

PC50135-1G PC50135-250MG
76606-174EA 619.34 USD
76606-174 76604-110
2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%
2-Bromo-4-chloro-6-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD24848700
CAS Number: 1449008-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR