4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC500654-1G PC500654-250MG
76626-668EA 846.74 USD
76626-668 76626-670
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile
Formula: C₁₀H₅F₆NO₂
Storage Temperature: Ambient
MDL Number: MFCD28103422
CAS Number: 1845713-67-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR