3-Bromo-6-chloro-2-fluorobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Bromo-6-chloro-2-fluorophenyl)methanol

PC500497-10G PC500497-1G PC500497-5G
76607-524EA 956.52 USD
76607-524 76603-964 76607-754
3-Bromo-6-chloro-2-fluorobenzyl alcohol 98%
3-Bromo-6-chloro-2-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD18447606
CAS Number: 886615-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR