3-Fluoro-4-iodobenzotrifluoride
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC500389-5G
PC500389-1G
PC500389-25G
76606-652EA
399.32
USD
76606-652
76607-798
76605-962
3-Fluoro-4-iodobenzotrifluoride
3-Fluoro-4-iodobenzotrifluoride
Formula:
C₇H₃F₄I Storage Temperature: Ambient |
MDL Number:
MFCD07771984 CAS Number: 132554-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...