2-Amino-4-bromo-3-fluorobenzoic acid 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC500189-1G
PC500189-250MG
PC500189-5G
76605-524EA
80.48
USD
76605-524
76607-446
76604-070
2-Amino-4-bromo-3-fluorobenzoic acid 95%
2-Amino-4-bromo-3-fluorobenzoic acid
Formula:
C₇H₅BrFNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD27664889 CAS Number: 1416013-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...