4-Hydroxy-2,3,5-trifluorobenzoic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,3,5-Trifluoro-4-hydroxybenzoic acid
, 4-Carboxy-2,3,6-trifluorophenol
PC49652-250MG
PC49652-1G
76604-308EA
202
USD
76604-308
76607-368
4-Hydroxy-2,3,5-trifluorobenzoic acid
4-Hydroxy-2,3,5-trifluorobenzoic acid
Formula:
C₇H₃F₃O₃ MW: 192.092 g/mol Melting Pt: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458460 CAS Number: 156839-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...