5-Bromo-3-fluoro-2-nitrophenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC49464-1G PC49464-250MG PC49464-5G
76605-040EA 166.04 USD
76605-040 76605-616 76606-588
5-Bromo-3-fluoro-2-nitrophenylacetic acid
5-Bromo-3-fluoro-2-nitrophenylacetic acid
Formula: C₈H₅BrFNO₄
Storage Temperature: Ambient
MDL Number: MFCD28740371
CAS Number: 1805938-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR