1-Ethynyl-4-(trifluoromethoxy)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC5952-1G PC5952-5G
76614-368EA 51.96 USD
76614-368 76614-370
1-Ethynyl-4-(trifluoromethoxy)benzene
1-Ethynyl-4-(trifluoromethoxy)benzene
Formula: C₉H₅F₃O
Density: 1.215 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD03094333
CAS Number: 160542-02-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR