2-Fluoro-4-methylsulfonylphenol 95+%

Supplier: APOLLO SCIENTIFIC
Danger

PC7486-1G PC7486-250MG PC7486-5G
76608-586EA 66.22 USD
76608-586 76608-588 76608-590
2-Fluoro-4-methylsulfonylphenol 95+%
2-Fluoro-4-methylsulfonylphenol
Formula: C₇H₇FO₃S
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD09264549
CAS Number: 398456-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR