2-Chloro-6-(trifluoromethyl)nicotinamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5877-1G PC5877-25G PC5877-5G
76603-304EA 24.26 USD
76603-304 76603-306 76603-308
2-Chloro-6-(trifluoromethyl)nicotinamide 98%
2-Chloro-6-(trifluoromethyl)nicotinamide
Formula: C₇H₄ClF₃N₂O
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD01862659
CAS Number: 386704-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR