2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC501059-1G PC501059-250MG PC501059-5G
76604-008EA 357.56 USD
76604-008 76605-618 76607-106
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 95%
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine
Formula: C₇H₆BrF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD09263998
CAS Number: 157590-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR