Potassium trifluoro(prop-1-en-2-yl)borate 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Potassium Isopropenyltrifluoroborate

PC49166-25G PC49166-5G
76623-724EA 63.16 USD
76623-724 76623-726
Potassium trifluoro(prop-1-en-2-yl)borate 95%
Potassium trifluoro(prop-1-en-2-yl)borate
Formula: C3H5BF3K
Melting Pt: 274…280 °C
Storage Temperature: Ambient
MDL Number: MFCD11052654
CAS Number: 395083-14-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR