3-Amino-2-fluorobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC6187-1G PC6187-5G
76603-526EA 20.36 USD
76603-526 76603-528
3-Amino-2-fluorobenzoic acid 98%
3-Amino-2-fluorobenzoic acid
Formula: C7H6FNO2
Storage Temperature: Ambient
MDL Number: MFCD06797827
CAS Number: 914223-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR