2-Chloro-6-(trifluoromethyl)nicotinonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5448-1G PC5448-5G
76601-998EA 119.48 USD
76601-998 76602-000 76602-002
2-Chloro-6-(trifluoromethyl)nicotinonitrile 98%
2-Chloro-6-(trifluoromethyl)nicotinonitrile
Formula: C₇H₂ClF₃N₂
Boiling Pt: 74…76 °C/0.8mmHg
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD01862660
CAS Number: 386704-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR