5-(Trifluoromethyl)picolinic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC9132-1G PC9132-5G
76610-152EA 20.87 USD
76610-152 76610-154
5-(Trifluoromethyl)picolinic acid 97%
5-(Trifluoromethyl)picolinic acid
Formula: C₇H₄F₃NO₂
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD04113632
CAS Number: 80194-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR