3,4-Dinitrofluorobenzene 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-Fluoro-1,2-dinitrobenzene
PC3040D-25G
PC3040D-1G
PC3040D-5G
76618-144EA
130
USD
76618-144
76618-142
76618-146
3,4-Dinitrofluorobenzene 97%
3,4-Dinitrofluorobenzene
Formula:
C₆H₃FN₂O₄ MW: 186.1 g/mol Melting Pt: 55…56 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236671 CAS Number: 364-53-4 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...