Methyl-3-amino-4-fluorobenzoate 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3-amino-4-fluorobenzoate

PC7646-1G PC7646-25G PC7646-5G
76608-688EA 20.87 USD
76608-688 76608-690 76608-692
Methyl-3-amino-4-fluorobenzoate 99%
Methyl 3-amino-4-fluorobenzoate
Formula: C₈H₈FNO₂
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689743
CAS Number: 369-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR