5-Bromo-1,3-dichloro-2-fluorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dichloro-4-fluorobromobenzene

PC8928-100G PC8928-25G PC8928-5G
76619-012EA 61.12 USD
76619-012 76619-014 76619-016
5-Bromo-1,3-dichloro-2-fluorobenzene
5-Bromo-1,3-dichloro-2-fluorobenzene
Formula: C₆H₂BrCl₂F
Boiling Pt: 227 °C
Melting Pt: 22.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09996901
CAS Number: 17318-08-0
UN: 3082
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR