3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC4377J-10G
PC4377J-1G
PC4377J-5G
76626-500EA
111.06
USD
76626-500
76626-502
76626-504
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile
Formula:
C₉H₅F₄N Storage Temperature: Ambient |
MDL Number:
MFCD00061181 CAS Number: 239087-12-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...