3-Amino-4-fluorophenol 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC7511-25G PC7511-5G
76618-910EA 112.06 USD
76618-910 76618-912
3-Amino-4-fluorophenol 99%
3-Amino-4-fluorophenol
Formula: C₆H₆FNO
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD09033850
CAS Number: 62257-16-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR