3-Fluoro-2-formylphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid

PC32758-1G PC32758-25G PC32758-5G
76623-334EA 20.87 USD
76623-334 76623-336 76623-338
3-Fluoro-2-formylphenylboronic acid
3-Fluoro-2-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697421
CAS Number: 871126-15-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR