4'-Methyl-3'-(trifluoromethyl)acetophenone 97%

Supplier: APOLLO SCIENTIFIC

PC9188-10G PC9188-1G PC9188-5G
76610-200EA 686.58 USD
76610-200 76610-202 76610-204
4'-Methyl-3'-(trifluoromethyl)acetophenone 97%
4'-Methyl-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD08532499
CAS Number: 851262-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR