2-Bromo-3-(trifluoromethyl)benzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5274-1G PC5274-250MG
76600-674EA 35.66 USD
76600-674 76600-676
2-Bromo-3-(trifluoromethyl)benzoic acid 98%
2-Bromo-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD08532493
CAS Number: 177420-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR