4-Bromophenylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Bromo-4-(pentafluorosulfanyl)benzene, 1-Bromo-4-(pentafluorosulphanyl)benzene

PC8054-1G PC8054-5G
76618-948EA 172.16 USD
76618-948 76618-950 76618-952
4-Bromophenylsulfur pentafluoride
4-Bromophenylsulphur pentafluoride
Formula: C₆H₄BrF₅S
Boiling Pt: 77…78 °C/10mmHg
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425928
CAS Number: 774-93-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR