HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)HBTU

PC6587-1KG PC6587-500G PC6587-100G
76603-732EA 13.39 USD
76603-732 76603-734 76603-736
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

Peptide coupling reagent.

Formula: C₁₁H₁₆F₆N₅OP
MW: 379.2 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00075445
CAS Number: 94790-37-1
EINECS: 423-020-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR